A propos de cet article
These 25/42-place fusion furnaces are available with LPG, natural gas or diesel fuel source options. Crucibles should be 25 x 50g or 42 x 40g.
- Suitable for small labs; 100-400 samples per day including standards
- Base construction uses high-quality 75 x 75 RHS
- Furnace body construction is durable 5mm laser-cut plate
- High quality refractory brick and insulation
- Bolt holes in base plates of all 4 legs for individual adjustment on uneven surfaces
- Pneumatic door controls with foot pedal controller—available with an optional hand valve controller
This furnace has a fan forced single burner system with an automatic igniton system
The door has a pneumatic ram opening system with an easy to use foot pedal control or optional hand valve that allows you to open the door part-way so that you can check inside with minimal temperature loss.
Temperature is monitored and modulated automatically by a Type K thermocouple system and Omron temperature controller pre-set for a maximum temperature of 1200 degrees Celsius. This controller is simple to operate, and has a 2-level display (PV and SV) that shows actual temperature as well as the temperature the furnace is set to.
Spécifications
- Catalog No:
- CA11023-238
- CA11023-240
- CA76047-128
- CA76047-130
- CA11023-242
- Shipping dimensions:
- 151 x 169 x 208 cm
- 151 x 169 x 208 cm
- 151 x 169 x 208 cm
- 151 x 169 x 208 cm
- 151 x 169 x 208 cm
- Shipping weight:
- 1700 kg
- 1700 kg
- 1700 kg
- 1700 kg
- 1700 kg