Bead Plier, Long Needle Nose, Excelta
# de catalogue 94024-208
Fournisseur: Excelta
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Length:16.5 cm (6.5")
- Description:Long Needle Nose Bead Plier
- Material:Stainless steel
- Supplier No.:2910
Spécifications
A propos de cet article
Two star, heavy stainless steel needle nose pliers are ideal for picking up beads.
Customers Who Bought This Also Bought