ContactUsLinkComponent
Sash-Glider Kit, Labconco
Sash-Glider Kit, Labconco
# de catalogue 10127-516
Fournisseur:  Labconco
Product available on GSA Advantage®
Sash-glider kit, weight: 5. 0 lbs, weight metric: 2. 0 kg
Sash-Glider Kit, Labconco
# de catalogue 10127-516
Fournisseur:  Labconco
Product available on GSA Advantage®
Spécifications
  • Description:
    Sash-Glider Kit
  • Cat. no.:
    10127-516
  • Supplier No.:
    3408500
Spécifications
A propos de cet article
Use the Sash-Glider to clean both sides of a biological safety cabinet's sash while it remains closed, to avoid contamination risks.
Delivery: Sash-Glider Kit includes magnetic outside and inside blades, 2 microfiber sheets and 10 disposable white scrubbing strips. Additional scrubbing strips are available.
Customers Who Bought This Also Bought