Waste Systems, Canister Kits, S.C.A.T.
Fournisseur: S.C.A.T.
Certificats
A propos de cet article
Complete waste systems including canister, SafetyWasteCap and exhaust filter.
- Delivery is made complete, system is immediately ready for use
- Only one article number for the comprehensive safety system
- No time consuming compilation of a configuration
- Price advantage compared to ordering piece by piece
All kits provide small exhaust filter, 2× capillary connector with 2.3/3.2 mm Ext.Ø, and 1× tube connector 6.4 to 9.0 mm interior diameter. The 5 and 10 L canister kits come with a SafetyWasteCap GL45 and the Space-Saving Canister Kit comes with SafetyWasteCap S50.
Customers Who Bought This Also Bought