Safety Funnel Disposal Units, S.C.A.T.
Fournisseur: Avantor
Certificats
A propos de cet article
Sets offer a way to get your waste fluid collection site ready for operation quickly. Kits come with grounding cable, sieve, and are PE-HD Electrically Conductive. A 10L container that is also PE-HD Electricallly Conductive is included in each kit. Ball Valve Funnel Kits have spalsh protection. Floats are for level control.
- Only one part number for the comprehensive safety system
- No time consuming compilation of a configuration
- Price advantage compared to ordering piece by piece
- The delivery is made complete – the system is immediately ready for use
Customers Who Bought This Also Bought