Canisters with Integrated Floater, S.C.A.T.
Fournisseur: S.C.A.T.
Certificats
A propos de cet article
Safely collect fluid laboratory waste. Ideal for use with S.C.A.T safety funnels.
The integrated floater gives a warning in time to prevent overfilling. Container from electrically conductive plastic counteracts electrostatic discharge and can be grounded. Thus they are especially well-suited for use in protected areas. Viewing Strip option has UN-Y approval.
Customers Who Bought This Also Bought