Safety Funnel with Hinged Lid, S.C.A.T
Fournisseur: S.C.A.T.
Certificats
A propos de cet article
Sieve is for trapping stirrers or larger contaminant particles.
Now made of PE-HD instead of stainless steel, so no corrosion by acids or lyes. Hinged lid allows for a clean closure. Freely rotatable cap comes in different thread sizes for a wide variety of containers. Safety lance ensures clean outflow without contaminating the container opening. With electrically conductive funnels (black), the lance facilitates a safe additional grounding of the contents. Models in black are made of electrically conductive PE-HD and have a ground connection to prevent sparking hazards.
Customers Who Bought This Also Bought