SafeLock Valve Couplings, S.C.A.T.
Fournisseur: Avantor
Certificats
A propos de cet article
The quick closures allow safe and quick connection of tubing and make bottle changes even easier
The bottles are rapidly disconnected with a single click and can be refilled at a safe location. Thanks to the integrated valve function, the closed safety system is maintained even when a tube is disconnected
Customers Who Bought This Also Bought