Thermal-Fused Laminate Lockers with Integrated Base, Wisconsin Bench
Supplier: Wisconsin Bench (WB)
Certificates
About this item
Impact resistant thermal-fused laminate (TFL) lockers features a 0.125" reveal, nickel finish Blum European style clip hinge with 120 degree opening, adjustable in all planes of operation and right mount typical.
- Lockers available in tier one, two, three and five door configurations
- Locker sides, backs and partitions are constructed using 45 lbs. density industrial grade particle board core
- Two hinges are installed on doors up to 42"H, three hinges on doors that exceed 42"H
- Integrated 4" base with levelers in each corner which easily adjust from the inside
Laminate color is consistent inside and out and all exposed edges are finished with matching PVC edge banding consistent with all REplay constructed products from Wisconsin Bench Manufacturing. Factory determined lock placement, typically centered vertically along edge of door with consideration given to lock type, pull or knob style and other accessories.
Certifications: GREENGUARD gold certified.
Ordering information: Key captive lock with two keys per opening included. Tier 1 lockers have one shelf and 2 hooks; Tier 2 and Tier 3 lockers have one hook and Tier 5 have no hook. Shown in Wild Cherry TFL laminate but also available in Folkstone and Solar Oak. Ships fully assembled, LTL bulk packed. Dimensions given are as installed.
Specifications
- Catalog No:
- 10054-764
- 10054-762
- 10055-004
- 10054-950
- 10054-760
- 10055-002
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Environmental certification:
- GREENGUARD Gold Certified
- GREENGUARD Gold Certified
- GREENGUARD Gold Certified
- GREENGUARD Gold Certified
- GREENGUARD Gold Certified
- GREENGUARD Gold Certified