Contemporary Bench with ButcherBlock Top, TFL with 3 mm PVC Edge, Wisconsin Bench
Supplier: Wisconsin Bench (WB)
Certificates
About this item
1" thick thermal-fused laminate both sides, subtop and apron with particle board core and matching 3mm PVC edges with incorporated levelers in each corner
1.25" thick hardwood top has 2 coats top and 1 coat bottom clear catalyzed lacquer for long-lasting beauty and durability. Hardwood top has radius corners and edges for comfort.
Certifications: GREENGUARD Gold Certified.
Ordering information: Shown with Wild Cherry finish, but currently available only in Folkstone finish. Ships fully assembled, LTL bulk packed. Dimensions given are as installed.
Customers Who Bought This Also Bought