Contemporary Benches, TFL with 3 mm PVC Edge, Wisconsin Bench
Supplier: Wisconsin Bench (WB)
Certificates
About this item
Thermal-fused laminate locker bench with particle board core and matching 3 mm PVC edge.
- Available in 3 colors: folkstone, solar oak and wild cherry
- Includes incorporated levelers in each corner
Matching 1" thick top applied over subtop with center support apron.
Certifications: GREENGUARD gold certified.
Ordering information: Shown in wild cherry. Ships fully assembled, LTL bulk packed. Dimensions given are as installed.
Specifications
- Catalog No:
- 10054-720
- 10054-718
- 10054-724
- 10054-722
- 10054-930
- 10054-714
- 10054-712
- 10054-932
- 10054-716
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Environmental certification:
- GREENGUARD Gold Certified
- GREENGUARD Gold Certified
- GREENGUARD Gold Certified
- GREENGUARD Gold Certified
- GREENGUARD Gold Certified
- GREENGUARD Gold Certified
- GREENGUARD Gold Certified
- GREENGUARD Gold Certified
- GREENGUARD Gold Certified