Phenolic Bench Kits with Stainless Steel Pedestals, 12" Width, Wisconsin Bench
Supplier: Wisconsin Bench (WB)
Certificates
About this item
1" thick phenolic locker room bench with either 2, 3 or 4 Stainless Steel 16.25"H pedestals
12" phenolic benches are a perfect fit for any locker room with its high resistance to humidity, abrasions and chemicals and features eased edges and radius corners for comfort. Pedestals are 14-gauge T201 Stainless Steel with 8.25" plate top and bottom for support.
Ordering information: 12"W x 1" thick phenolic bench with 2 Stainless Steel pedestals for lengths of 48", 3 Stainless Steel pedestals for lengths of 72" and 96"; and 4 Stainless Steel pedestals for 120" length bench. Hardware to secure bench to pedestal are included and should be installed with no more than 4 feet span between pedestals. Hardware to mount kit to the floor are not included. Ships in multiple cartons. Dimensions given are as installed.
Frequently Bought Together