Ergonomic Shovel, Small Blade, Remco
# de catalogue 76008-476
Fournisseur: Remco Products
Spécifications
- Length:49"
- Color:White
- Material:PP
- Dimensions:11W×H"
- Weight:3.8 lbs.
Spécifications
A propos de cet article
This robust ergonomic shovel is designed for use in direct contact with food.
The handle is reinforced with an inner aluminum tube. It is stepped in two places which helps to prevent back injuries. The neck joint has been strengthened in order to withstand heavy duty use. The long and small blade combination makes it suitable for shoveling smaller volumes and reaching into deep containers.
Certifications: FDA compliant raw material.
Customers Who Bought This Also Bought