Versilon™ C-219-A Flexible Chemical Transfer Tubing
Supplier: Saint Gobain Life Sciences
Certificates
About this item
General use clear vinyl tubing is supplied in 30.5 m (100') lengths.
Specifications
- Catalog No:
- 63009-365
- 63009-375
- 63009-355
- 63009-363
- 63009-351
- 63009-353
- 63009-346
- 63009-348
- 63009-404
- 63009-344
- 63009-400
- Durometer hardness:
- Shore A, 67
- Shore A, 67
- Shore A, 67
- Shore A, 67
- Shore A, 67
- Shore A, 67
- Shore A, 67
- Shore A, 67
- Shore A, 67
- Shore A, 67
- Shore A, 67