Dust Pan, Remco
# de catalogue 76006-992
Fournisseur: Remco Products
Spécifications
- Color:Black
- Material:PP
- Cat. no.:76006-992
- Length:12"
- Weight:0.4 lbs.
Spécifications
A propos de cet article
This non-food grade dustpan is a great alternative to more costly hygienic dustpans
It helps clean up after spills to ensure a clean working area.
Customers Who Bought This Also Bought