Dual Thread Adapter, Remco Products
# de catalogue 76006-982
Fournisseur: Remco Products
Spécifications
- Color:White
- Material:PP
- Weight:0.1 lbs.
Spécifications
A propos de cet article
This dual thread adapter allows Remco’s safety shovel blades to be used in conjunction with any European-thread handle
It affords Remco’s safety tools a higher degree of flexibility to meet the needs of a variety of safety tasks.
Customers Who Bought This Also Bought