ContactUsLinkComponent
Dual Thread Adapter, Remco Products
Dual Thread Adapter, Remco Products
# de catalogue 76006-982
Fournisseur:  Remco Products
Dual Thread Adapter 5.4 in
Dual Thread Adapter, Remco Products
# de catalogue 76006-982
Fournisseur:  Remco Products
Spécifications
  • Color:
    White
  • Material:
    PP
  • Weight:
    0.1 lbs.
Spécifications
A propos de cet article
This dual thread adapter allows Remco’s safety shovel blades to be used in conjunction with any European-thread handle
It affords Remco’s safety tools a higher degree of flexibility to meet the needs of a variety of safety tasks.
Customers Who Bought This Also Bought