Spécifications
- Dimensions:90 x 25 mm
- Description:High Profile, Semi-Stacking Petri Dish without Vents
- Cat. no.:CA73370-024
Spécifications
A propos de cet article
These high-profile Petri dishes are specifically designed to accommodate more media.
- Ideal for longer culture periods
- Semi-stacking
- Clear
They are excellent for mycology, histology and pathology labs, as well as seed germination work. These dishes are ideal when longer culture periods are required.