VWR® Planton Plant Tissue Culture Container
# de catalogue 10081-038
Fournisseur: VWR International
Spécifications
- Width:7.5 cm
- Description:Plant Tissue Culture Container
- Height:10 cm
- Cat. no.:10081-038
- Depth:7.5 cm
Spécifications
A propos de cet article
This autoclavable container features a transparent polycarbonate body and a polypropylene cap.
- Transparent
- Polycarbonate body and polypropylene cap construction
- Unique design for plant tissue culture
Unique cap and drip ring design makes the container ideal for plant tissue culture.
Customers Who Bought This Also Bought