Furnace Scraper, Steel
# de catalogue CA11017-662
Fournisseur: Webber's Welding
Spécifications
- Description:Furnace scraper, drum size: 117 cm (46")
- Cat. no.:CA11017-662
- Supplier No.:FA127
Spécifications
A propos de cet article
The furnace scraper is ideal for spreading bone ash and scraping spills from the furnace
The durable handle is made from 6.3mm steel rod and the specially designed serrated blade (178 x 63.5 x 4.8mm mild steel) provides maximum scraping ability.
Ordering information: Contact your Avantor sales representative for more information.
Customers Who Bought This Also Bought