Cupel Fork, Webber's Welding
Fournisseur: Webber's Welding
Certificats
A propos de cet article
Cupel Fork is specially designed for heavy-duty applications against heat from ovens or furnaces, while its mild steel prongs provide durable and dependable service.
Ordering information: Overall length: 57"
Handle length: 46"
Handle length: 46"
Customers Who Bought This Also Bought