Crucible Loading Fork, Webber's Welding
Fournisseur: Webber's Welding
Certificats
A propos de cet article
Crucible Fork is specially designed for heavy-duty applications with user safety in mind.
- Aluminum pipe handle helps insulate against heat from ovens or furnaces
- Mild steel crucible holders provide durable and dependable service
Ordering information: Overall Length: 50"
Handle Length: 46"
Handle Length: 46"
Customers Who Bought This Also Bought