ContactUsLinkComponent
F.C. Bond Ball Mill, Bico
F.C. Bond Ball Mill, Bico
# de catalogue 20058-110
Fournisseur:  Bico
BALLMILL 12INDRUM 3PH 220 400V
F.C. Bond Ball Mill, Bico
# de catalogue 20058-110
Fournisseur:  Bico
Spécifications
  • Description:
    3 Phase, 60 Cycle
  • Cat. no.:
    20058-110
  • Electrical:
    220/440 V
  • Drum Size:
    12×12"
  • Supplier No.:
    395-50
Spécifications
A propos de cet article
Designed by F.C. Bond for use by cement companies and quality control laboratories throughout the world. Can be used in calculating the grindability of all ore.
Ordering information: 285 Ball Charge. Contact your Avantor sales representative for more information.
Customers Who Bought This Also Bought