VWR® Cleanroom Paper
Fournisseur: VWR International
Certificats
A propos de cet article
The cleanroom paper is ideal for documentation in critical environments, where very low particle and ESD generation is important.
- Low particle generation
Cleanroom paper is provided in latex or latex-free variants in a variety of colors.
Customers Who Bought This Also Bought