VWR® Traceable® Digital Humidity Temperature/Dew Point Meter | Avantor
VWR® Traceable® Digital Humidity Temperature/Dew Point Meter
Digital Humidity Temperature/Dew Point Meter, 2 5/8 × 3 7/8 × 5/8", –10…+60 °C (14...140 °F), ±1 °C, 6 oz.
Catalog # 89500-398
Supplier:   VWR International
VWR® Traceable® Digital Humidity Temperature/Dew Point Meter
Catalog # 89500-398
Supplier:  VWR International
VWR® Traceable® Digital Humidity Temperature/Dew Point Meter
VWR® Traceable® Digital Humidity Temperature/Dew Point Meter
Digital Humidity Temperature/Dew Point Meter, 2 5/8 × 3 7/8 × 5/8", –10…+60 °C (14...140 °F), ±1 °C, 6 oz.
Price
$238.09
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Each
Product Details & Documents
Convenient triple memories recall minimum/maximum readouts for relative humidity, dew point, and temperature.
  • Simultaneously displays all three current readings
  • Sits anywhere in the lab and updates every 30 seconds
  • Relative humidity range: 1 to 99%
  • Resolution: 0.1%
  • Accuracy: ±3% mid-range, ±5% elsewhere
  • Resolution: 0.1°
Simultaneously displays all three current readings. Always-on feature allows for monitoring conditions all night, over weekends, and for extended time periods. Audible and visual LED alarm may be set to trigger at three different humidity readings (55%, 60% or 65%). Solid state sensors deliver response times of less than 30 seconds. Oversized display supplies updates once a second.
Spécifications
  • Plage de température:
    –10…+60 °C (14...140 °F)
  • Désignation:
    Digital Humidity Temperature/Dew Point Meter
  • Dimensions:
    2 5/8 × 3 7/8 × 5/8"
  • Temperature Accuracy:
    ±1 °C
  • Weight:
    6 oz.