VWR® Dispenser Replacement Reservoir Bottles
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Low-profile reservoir reservoir bottle is designed for use with bottle top dispensers and bottle top burets.
- Amber borosilicate glass construction
- Sold individually
Amber borosilicate glass construction is ideal for light sensitive samples. Can be used with Acurex™ 501 Compact Bottle-Top Dispensers or VWR 501 Dispenser.
Customers Who Bought This Also Bought