Bottle, with Sealer Cap, LDPE, Dynalon
Fournisseur: Dynalon
Certificats
A propos de cet article
These low density polyethylene bottles are supplied with a sealer cap that snaps on easily after use and seals tightly.
Spout openings are standard 0.030", and can be enlarged by cutting back with scissors.
Customers Who Bought This Also Bought