Graduated Burettes, Acrylic, Dynalon
Fournisseur: Dynalon
Certificats
A propos de cet article
Break-resistant, transparent acrylic burettes are screen printed in black to Class B accuracy
They are suitable for use in education, research, or industrial laboratories, especially where breakages often occur. This series is unaffected by most dilute aqueous solutions (except ammonia and HF), but unsuitable for ethanol or organic solvents. Lengths and internal dimensions can vary as all are individually calibrated.
Customers Who Bought This Also Bought