Laboratory Bottles, Wide Mouth, LDPE, Bulk Packed, Dynalon
: Dynalon
Translucent, low density polyethylene wide mouth bottles are supplied with leakproof polypropylene screw closures.
Manufactured from FDA approved resin for use with food products. The LDPE construction offers greater flexibility and impact resistance than HDPE bottles. Bulk packed with closures included but not attached.
Customers Who Bought This Also Bought