Wash Bottles, Wide Neck, Printed, Dynalon
Supplier: Dynalon
Certificates
About this item
High-quality wash bottles are manufactured from flexible natural polyethylene and fitted with white, leak-proof, wide neck polypropylene spout closures.
- All vessels feature S.P.I. recycling codes on the base
- Maximum operating temperature of LDPE is 80 °C (176 °F)
The wide neck offers easy spill-free filling, and the acute angled, precision-molded dispensing spout ensures a consistent downward pointing jet.
Delivery: The wash bottles are supplied in shrink-wrapped packs of five.
Customers Who Bought This Also Bought