Laboratory Bottles, Wide Mouth, Polypropylene, Dynalon
Supplier: Dynalon
Certificates
About this item
Leakproof, rigid, and translucent wide mouth polypropylene lab bottles can be autoclaved.
- Excellent chemical resistance
- Ideal for storage and sampling of liquids or solids
Delivery: Supplied with a polypropylene closure.
Customers Who Bought This Also Bought