VWR® Volumetric Flasks without Stopper, Amber Glass, Class A, Serialized
Fournisseur: VWR International
Certificats
A propos de cet article
Made from borosilicate 3.3 expansion glass, flasks are serialized and supplied without stopper.
- Borosilicate glass construction
- Individually serialized
- Comes without a stopper
Flasks are made with a graduation mark for accurate measurements.
Ordering information: Flasks come with individual certificate of compliance.
Customers Who Bought This Also Bought