Bottle with Handle, HDPE, Dynalon
Supplier: Dynalon
Certificates
About this item
Lightweight HDPE bottle features a handle for convenient carrying.
- Bottles are not autoclavable
HDPE is resistant to most chemicals and will perform in conditions up to 120 °C (248 °F).
Delivery: Bottles are supplied with a 38 mm polypropylene screw closure with foam liner for tight seal.
Customers Who Bought This Also Bought