Squeegees, 28" Fixed Head Double Blade With Closed Cell Foam Refill Cassette, Remco Products
Fournisseur: Remco Products
Certificats
A propos de cet article
The neoprene rubber blades on this squeegee make it the most effective squeegee in the Vikan range
It is excellent on all types of floors, as the soft rubber reaches into and cleans all spaces on the floor, such as grouting between tiles. The cartridge system blades can be removed for easy cleaning and replaced when worn out, making the model a very economical option. Polypropylene material. Foam rubber blade material.
Customers Who Bought This Also Bought