A propos de cet article
The Valor® 4000 water-resistant food scale sets a new industry standard with the winning combination of touchless operations, speed, and durability
These compact bench scales are easy to clean and easy to transport. They provide a stabilization time of less than 0.5s, and feature a power jack auto-closed design. Additional standard features include a level bubble, fixed weighing pan, overload and underload indicators, low battery indicator, AC adapter, rechargeable battery, and a touchless sensor for Tare functions.
These rapid-response food scales enhance productivity in harsh environments.
Available in both stainless steel and plastic housing designs, the Valor® 4000 offers large LED front and rear displays, food safety and legal for trade approvals, and puncture resistant keypads. All features that combine to make it the premiere scale for use in dry or washdown environments.
Spécifications
- Catalog No:
- 10123-922
- 10123-926
- 10123-924
- 10123-898
- 10123-896
- 10123-894
- 10123-920
- 10123-900
- Operating temperature range:
- –10 to 40°C (14 to 104°F)
- –10 to 40°C (14 to 104°F)
- –10 to 40°C (14 to 104°F)
- –10 to 40°C (14 to 104°F)
- –10 to 40°C (14 to 104°F)
- –10 to 40°C (14 to 104°F)
- –10 to 40°C (14 to 104°F)
- –10 to 40°C (14 to 104°F)
- Voltage:
- 12V DC 0.84A, AC Universal Adapter (100-220V/50-60Hz) and Built-In Internal Rechargeable Lead Acid Battery Included
- 12V DC 0.84A, AC Universal Adapter (100-220V/50-60Hz) and Built-In Internal Rechargeable Lead Acid Battery Included
- 12V DC 0.84A, AC Universal Adapter (100-220V/50-60Hz) and Built-In Internal Rechargeable Lead Acid Battery Included
- 12V DC 0.84A, AC Universal Adapter (100-220V/50-60Hz) and Built-In Internal Rechargeable Lead Acid Battery Included
- 12V DC 0.84A, AC Universal Adapter (100-220V/50-60Hz) and Built-In Internal Rechargeable Lead Acid Battery Included
- 12V DC 0.84A, AC Universal Adapter (100-220V/50-60Hz) and Built-In Internal Rechargeable Lead Acid Battery Included
- 12V DC 0.84A, AC Universal Adapter (100-220V/50-60Hz) and Built-In Internal Rechargeable Lead Acid Battery Included
- 12V DC 0.84A, AC Universal Adapter (100-220V/50-60Hz) and Built-In Internal Rechargeable Lead Acid Battery Included