SP Bel-Art Drying Tubes with Tube Fittings, Bel-Art Products, a part of SP
Fournisseur: Bel-Art Products, a Part of SP
Certificats
A propos de cet article
Drying tubes with tube fittings can be filled with desiccant of choice.
- Polyethylene construction
- Virtually unbreakable
Each unbreakable polyethylene straight tubes, 16 mm int.Ø. and 19 mm ext.Ø. (⅝×¾"), is supplied with two polypropylene fittings with serrated tapered end that accept 6.4 to 9.5 mm (¼ to ⅜") int.Ø. tubing. Drying agent not included.
Customers Who Bought This Also Bought