GOLD BRAND API High Precision Combined Form Hydrometer, Thermco
Fournisseur: Thermco
Certificats
A propos de cet article
High precison API combined form hydrometers made to exceed API specifications, which incorporate a thermometer in the hydrometer
The temperature standardization is 60/60F. The hydrometer scale division are 2.0 degrees farenheit. The hydrometers will perform with the same accuracy as the ASTM hydrometers with mercury thermometers.
The thermometer is made with a non-mercury, blue spirit, has a range of 0 to 150 degrees farenheit and is mounted in thebody of the hydrometer. Length of hydrometer is 15 inches.
Used to measure the density according to API degree scale for petroleum applications.
Certifications: ASTM approved/ISO17025/Statement of Compliance and Traceability.
Customers Who Bought This Also Bought