PB-900™ Programmable Jar Tester, Phipps & Bird
Fournisseur: Phipps & Bird
Certificats
A propos de cet article
The Phipps & Bird Jar Tester has been re-engineered to improve its performance and bring it into compliance with CE and cTUVus requirements
The motor has been improved to a stepper motor drive system. Additional features include LED lighting modules and complete 24VDC with included adapter that will power the machine from 100 to 240V sources.
Customers Who Bought This Also Bought