Vikan® Deck/Wall Scrubs, Stiff, Remco Products
Fournisseur: Remco Products
Certificats
A propos de cet article
This wide scrubbing broom is ideal for cleaning heavily soiled areas and covering a lot of ground in food production facilities such as meat processing plants, bakeries, canneries, and dairy plants.
- Block material is polypropylene; bristle material is polyester
- Maximum temperature: 250°F
- Dishwasher safe
Certifications: FDA compliant raw material.
Customers Who Bought This Also Bought