Vikan® Waterfed Washing Brushes, Remco Products
Fournisseur: Remco Products
Certificats
A propos de cet article
This unique brush has split fiber bristles to absorb liquid so that the user is able to efficiently carry soapy water from bucket to the surface to be cleaned.
- Bristles are soft enough for sensitive surfaces
- Waterfed design makes it excellent for rinsing after foaming
- Neck angle is adjustable to allow all bristles to remain constantly in contact with the cleaning surface
- Autoclavable up to 250° F
Block material is polypropylene; bristle material is polyester.
Also commonly used as a dusting brush.
Width: 2.75''
Height: 5.5''
Certifications: FDA compliant raw material.
Customers Who Bought This Also Bought