Vikan® Deck/Wall Scrubs, Stiff, Remco Products
Fournisseur: Remco Products
Certificats
A propos de cet article
This floor scrubbing broom has firm bristles.
- Block material is polypropylene; bristle material is polyester
- Maximum temperature: 250°F
- Dishwasher safe
- Available in a variety of colors
The bristle length is perfect for scrubbing all types of floors; epoxy, tiles, cement, etc. It also reaches well into holes and grouting to remove hidden dirt.
Certifications: FDA compliant raw material.
Customers Who Bought This Also Bought