Vikan® High-Low Brushes / Scrubbing Brooms, Remco Products
Fournisseur: Remco Products
Certificats
A propos de cet article
The angled shape of this scrubbing broom was designed to clean the area where the wall meets the floor, but also effective for cleaning the floor itself.
- Block material is polypropylene; bristle material is polyester
- Dishwasher safe
- Autoclavable up to 250°F
Its bristles are gentle enough to be used on equipment. It is a popular choice in meat processing facilities, canneries, mills, wineries and breweries—and generally any moist environment that creates difficult cleaning challenges
Certifications: FDA compliant raw material.
Customers Who Bought This Also Bought