Vikan® Tank Brushes, Medium, Remco Products
Fournisseur: Remco Products
Certificats
A propos de cet article
Ideal for cleaning tanks and vats, and is also suitable for cleaning drain gulleys.
- Block material is polypropylene; bristle material is polypropylene
- Dishwasher safe
- Autoclavable up to 250 °F
May be used with any European-thread handle from Vikan.
Certifications: FDA compliant raw material.
Customers Who Bought This Also Bought