GOLD BRAND ASTM/API High Precision Plain Form Hydrometer, Thermco
Fournisseur: Thermco
Certificats
A propos de cet article
High precison ASTM plain form hydrometers made to exceed ASTM and API specifications, which do not incorporate a thermometer in the hydrometer
The temperature standardization is 60/60F. The hydrometer scale division are 0.1 degree.
Tall form, 330 mm, has 12 degree ranges; Short form, 163 mm, has 6 degree ranges.
Used to measure the density according to API degree scale for petroleum applications.
Certifications: ASTM approved/ISO17025/Statement of Compliance and Traceability.
Ordering information: Hard square plastic tube included.
Spécifications
- Catalog No:
- 10053-072
- 10053-074
- 10053-076
- 10053-078
- 10053-080
- 10053-082
- 10053-084
- 10053-086
- 10053-046
- 10053-048
- 10053-090
- 10053-050
- 10053-052
- 10053-054
- 10053-056
- 10053-058
- 10053-102
- 10053-060
- 10053-104
- 10053-062
- 10053-064
- 10053-066
- 10053-068
- 10053-070
- Environmentally Preferable:
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient
- Energy Efficient