ASTM-Approved Non-Mercury Tank Gauging Thermometers, Thermco
Fournisseur: Thermco
Certificats
A propos de cet article
Wood Brass Cup model, case made from a hardwood that prevents absorbtion of the petroleum sample
Thermometers supplied with each case are ASTM approved, non-mercury blue spirit; remounted on spring clips and can be replaced if breakage occurs. Three ranges available in both celcius and farenheit.
The attached brass cup is non-sparking and meets ASTM and API requirements.
Certifications: ASTM approved/ISO17025/Statement of Traceability.
Ordering information: Woodback Cup Case Thermometer Only. Case is available separately.
Spécifications
- Catalog No:
- 10053-482
- 10053-036
- 10053-038
- 10053-040
- 10053-042
- 10053-480
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health