ASTM Approved Non-Mercury Tank Gauging Thermometers, Thermco
Fournisseur: Thermco
Certificats
A propos de cet article
Scoop Master model, a one piece molded black heat resistant plastic material impervious to the chemical action of petroleum products and aromatic hydrocarbons
Thermometers supplied with each case are ASTM approved, non-mercury blue spirit; remounted on spring clips and can be replaced if breakage occurs. Three ranges available in both celcius and farenheit.
The low thermal conductivity of the plastic maintains the temperatures of the sample. The case is non-sparking.
Certifications: ASTM approved/ISO17025/Statement of Traceability.
Ordering information: Cup Case Thermometer Only. Case is available separately.
Spécifications
- Catalog No:
- 10053-466
- 10053-468
- 10053-470
- 10053-472
- 10053-474
- 10053-476
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health