ASTM Approved Non-Mercury Thermometers, Thermco
Fournisseur: Thermco
Certificats
A propos de cet article
ASTM approved non-mercury blue spirit "precision" thermometers with the same specifications as the mercury ASTM thermometers
Each is designated for an ASTM method, serialized, filled with a blue non-mercury liquid with a white background for easy reading of temperature values.
Certifications: ASTM approved/ISO17025/Statement of Traceability.
Ordering information: Hard plastic square tube included.
Spécifications
- Catalog No:
- 10052-992
- 10052-994
- 10052-996
- 10052-998
- 10053-000
- 10053-002
- 10053-228
- 10053-004
- 10053-230
- 10053-006
- 10053-232
- 10053-008
- 10053-234
- 10053-010
- 10053-236
- 10052-990
- 10053-238
- 10053-240
- 10053-242
- 10053-012
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health