Order Entry
ContactUsLinkComponent
Human Recombinant BMP3 (from E. coli)
Human Recombinant BMP3 (from E. coli)
# de catalogue 10073-014
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant BMP3 (from E. coli)
# de catalogue 10073-014
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Size:
    0.01 mg
  • Biological activity:
    Biological activity was determined by its ability to inhibit BMP-2-induced alkaline phosphatase production by ATDC-5 cells. Endotoxin level is less than 0.1 ng per μg (1EU/μg).
  • Protein synonyms:
    BMP3A|BMP-3A|Bone morphogenetic protein 3A|Bone morphogenetic protein 3|BMP-3
  • Protein/peptide name:
    BMP3
  • Purity:
    >95%
  • Formulation:
    Lyophilized
  • Cat. no.:
    10073-014
  • Supplier No.:
    40-547

Spécifications

A propos de cet article

TGF-β family members are key modulators of cell proliferation, differentiation, matrix synthesis, and apoptosis. As implied by their name, BMPs initiate, promote, and regulate the development, growth and remodeling of bone and cartilage. In addition to this role, BMPs are also involved in prenatal development and postnatal growth, remodeling and maintenance of a variety of other tissues and organs. BMP-3 is abundantly found in adult bone, and to a lesser extent fetal cartilage. BMP-3 inhibits osteogenesis and bone formation by activating a signaling cascade that antagonizes the signaling of pro-osteogenic BMPs. Recombinant human BMP-3 is a disulfide linked homodimeric protein that corresponds to residues 361 to 472 of the 472 amino acid BMP-3 precursor protein.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought