Order Entry
ContactUsLinkComponent
Human Recombinant Noggin (from HEK293 Cells)
Human Recombinant Noggin (from HEK293 Cells)
# de catalogue 10072-758
Fournisseur:  Prosci
Numéro CAS:  
Human Recombinant Noggin (from HEK293 Cells)
# de catalogue 10072-758
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Size:
    0.005 mg
  • Biological activity:
    Biological activity was determined by its ability to inhibit 5.0 ng/mL of BMP - 4 induced alkaline phosphatase production by ATDC chondrogenic cells. The expected ED50 for this effect is 2.0 - 3.0 ng/mL of Noggin.
  • Protein synonyms:
    SYM1|SYNS1|Noggin
  • Protein/peptide name:
    Noggin
  • Formulation:
    Lyophilized
  • Cat. no.:
    10072-758
  • Supplier No.:
    40-207

Spécifications

A propos de cet article

Noggin belongs to a group of diffusible proteins which bind to ligands of the TGF-β family and regulate their activity by inhibiting their access to signaling receptors. The interplay between TGF-β ligands and their natural antagonists has major biological significance during development processes, in which cellular response can vary considerably depending upon the local concentration of the signaling molecule. Noggin was originally identified as a BMP-4 antagonist whose action is critical for proper formation of the head and other dorsal structures. Consequently, Noggin has been shown to modulate the activities of other BMPs including BMP-2,-7,-13, and -14. Targeted deletion of Noggin in mice results in prenatal death and recessive phenotype displaying a severely malformed skeletal system. Conversely, transgenic mice over-expressing Noggin in mature osteoblasts display impaired osteoblastic differentiation, reduced bone formation, and severe osteoporosis. Recombinant human Noggin is a 46 kDa disulfide-linked homodimer (120-10C) consisting of two 206 amino acid polypeptide chains. Monomeric glycosylated Noggin migrates at an apparent molecular weight of approximately 28.0-33.0 kDa by SDS PAGE analysis under reducing conditions.

Store in working aliquots at -20°C. Avoid freeze/thaw cycles.

Customers Who Bought This Also Bought