Paddle Scrapers, 40" with Holes, Remco Products
Fournisseur: Remco Products
Certificats
A propos de cet article
This two-in-one tool is handy for both mixing and scraping, and is great for a variety of applications such as soup, confectionery, dough, and sauce manufacturing as well as many other food processing or other mixing tasks
One-piece construction and the unique long-handle design make it easy to use for many jobs. Plastic is a lightweight alternative to metal mixing paddle scrapers, and will reduce fatigue for the user. Polypropylene material.
Certifications: FDA compliant raw material.
Customers Who Bought This Also Bought