Order Entry
ContactUsLinkComponent
Jejunum Lysate, ProSci
Jejunum Lysate, ProSci
# de catalogue 10062-190
Fournisseur:  Prosci
Numéro CAS:  
Jejunum Lysate, ProSci
# de catalogue 10062-190
Fournisseur:  Prosci
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Cell Format:
    Cryopreserved
  • Description:
    Normal
  • Size:
    0.1 mg
  • Storage conditions:
    Store at 2-8°C for continuous use. For extended storage, freeze working aliquots at -70°C.
  • Storage buffer:
    Jejunum lysate is supplied in SDS sample buffer containing 5% β-mercaptoethanol.
  • Concentration:
    2 mg/mL
  • Tested applications:
    WB
  • Cat. no.:
    10062-190
  • Source:
    Human
  • Tissue:
    Jejunum
  • Supplier No.:
    1368

Spécifications

A propos de cet article

Jejunum tissue lysate was prepared by homogenization in lysis buffer (10 mM HEPES pH7.9, 1.5 mM MgCl2, 10 mM KCl, 1 mM ethylenediaminetetraacetic acid, 10% glycerol, 1% NP-40, and a cocktail of protease inhibitors)

Tissue and cell debris was removed by centrifugation. The product was boiled for 5 min in 1 x SDS sample buffer (50 mM Tris-HCl pH 6.8, 12.5% glycerol, 1% sodium dodecylsulfate, 0.01% bromophenol blue) containing 5% β-mercaptoethanol.

Customers Who Bought This Also Bought